Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NEPRO
Entrez Gene ID
102183266
Gene Name
nucleolus and neural progenitor protein
Gene Aliases
-
Chromosome Location
Chr.1: 57487946 - 57506500 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005674925.3 XP_005674982.2 3 - 4 410 76
XM_013962566.2 XP_013818020.2 2 - 3 249 76
XM_018066177.1 XP_017921666.1 3 - 4 275 76
XM_018066237.1 XP_017921726.1 3 - 4 270 76

View Full Product Details