Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NEPRO
Entrez Gene ID
102183266
Gene Name
nucleolus and neural progenitor protein
Gene Aliases
-
Chromosome Location
Chr.1: 57487946 - 57506500 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005674925.3 XP_005674982.2 4 - 5 546 93
XM_013962566.2 XP_013818020.2 3 - 4 385 93
XM_018066177.1 XP_017921666.1 4 - 5 411 93
XM_018066237.1 XP_017921726.1 4 - 5 406 93
XM_018066416.1 XP_017921905.1 1 - 2 106 93

View Full Product Details