Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC102186369
Entrez Gene ID
102186369
Gene Name
claudin-22
Gene Aliases
CLDN22
Chromosome Location
Chr.27: 31383095 - 31383767 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005698750.3 XP_005698807.3 1 - 1 625 71

View Full Product Details