Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAML1
Entrez Gene ID
102181584
Gene Name
mastermind like transcriptional coactivator 1
Gene Aliases
-
Chromosome Location
Chr.7: 107200234 - 107261850 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018051602.1 XP_017907091.1 2 - 3 2030 72

View Full Product Details