Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NTSR1
Entrez Gene ID
102184259
Gene Name
neurotensin receptor 1
Gene Aliases
-
Chromosome Location
Chr.13: 53929717 - 53977829 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005688283.3 XP_005688340.2 2 - 3 919 92

View Full Product Details