Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HCK
Entrez Gene ID
102183977
Gene Name
HCK proto-oncogene, Src family tyrosine kinase
Gene Aliases
-
Chromosome Location
Chr.13: 60870317 - 60915477 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018057690.1 XP_017913179.1 3 - 4 462 89
GenBank mRNA JO610655.2 - - 1795 89

View Full Product Details