Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB38
Entrez Gene ID
102183040
Gene Name
RAB38, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.29: 6931754 - 6998335 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005699431.3 XP_005699488.1 1 - 2 339 112
GenBank mRNA KA323195.1 - 1 - 2 391 112

View Full Product Details