Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZFPL1
Entrez Gene ID
102188020
Gene Name
zinc finger protein like 1
Gene Aliases
-
Chromosome Location
Chr.29: 43722602 - 43728652 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005699878.3 XP_005699935.1 4 - 5 598 103
XM_018043488.1 XP_017898977.1 4 - 5 600 103
XM_018043489.1 XP_017898978.1 4 - 5 540 103
XM_018043490.1 XP_017898979.1 4 - 5 598 103

View Full Product Details