Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ALX3
Entrez Gene ID
102184773
Gene Name
ALX homeobox 3
Gene Aliases
-
Chromosome Location
Chr.3: 87201957 - 87212192 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018045887.1 XP_017901376.1 2 - 3 673 60

View Full Product Details