Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OLFML2A
Entrez Gene ID
102183077
Gene Name
olfactomedin like 2A
Gene Aliases
-
Chromosome Location
Chr.11: 95296800 - 95324120 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA JT701640.1 - 7 - 8 987 65

View Full Product Details