Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- NXPH1
- Entrez Gene ID
-
102172321
- Gene Name
- neurexophilin 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.4: 103758868 - 104122536 on Build ARS1
- Gene Symbol
- GGT7
- Entrez Gene ID
-
102175220
- Gene Name
- gamma-glutamyltransferase 7
- Gene Aliases
-
-
- Chromosome Location
-
Chr.13: 63732341 - 63756691 on Build ARS1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
JO644073.2 |
- |
- |
2922 |
73 |
View Full Product Details