Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VSNL1
Entrez Gene ID
102179016
Gene Name
visinin like 1
Gene Aliases
-
Chromosome Location
Chr.11: 80666421 - 80791692 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005686987.3 XP_005687044.1 3 - 4 795 59

View Full Product Details