Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TFF2
Entrez Gene ID
102182174
Gene Name
trefoil factor 2
Gene Aliases
-
Chromosome Location
Chr.1: 142195364 - 142198458 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005675636.3 XP_005675693.1 1 - 2 130 95

View Full Product Details