Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BAAT
Entrez Gene ID
102174570
Gene Name
bile acid-CoA:amino acid N-acyltransferase
Gene Aliases
-
Chromosome Location
Chr.8: 90845021 - 90861304 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005684245.3 XP_005684302.1 2 - 3 592 61

View Full Product Details