Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HHLA1
Entrez Gene ID
102169030
Gene Name
HERV-H LTR-associating 1
Gene Aliases
-
Chromosome Location
Chr.14: 72906273 - 72931261 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005688938.2 XP_005688995.1 3 - 4 198 88

View Full Product Details