Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZSCAN29
Entrez Gene ID
102180633
Gene Name
zinc finger and SCAN domain containing 29
Gene Aliases
-
Chromosome Location
Chr.21: 54617439 - 54628212 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005695293.3 XP_005695350.2 2 - 3 535 70

View Full Product Details