Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LY6G5C
Entrez Gene ID
102179997
Gene Name
lymphocyte antigen 6 family member G5C
Gene Aliases
-
Chromosome Location
Chr.23: 22339075 - 22341507 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005696665.2 XP_005696722.1 2 - 3 183 67

View Full Product Details