Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VMA21
Entrez Gene ID
102170497
Gene Name
vacuolar ATPase assembly factor VMA21
Gene Aliases
-
Chromosome Location
Chr.Un|NW_017189518: 400672 - 411366 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005700483.3 XP_005700540.1 2 - 3 297 105
XM_018044322.1 XP_017899811.1 3 - 4 557 105
XM_018044323.1 XP_017899812.1 3 - 4 506 105

View Full Product Details