Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CETN3
Entrez Gene ID
102177795
Gene Name
centrin 3
Gene Aliases
-
Chromosome Location
Chr.7: 20573157 - 20591120 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005683427.3 XP_005683484.1 2 - 3 295 71

View Full Product Details