Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MCHR2
Entrez Gene ID
102169386
Gene Name
melanin concentrating hormone receptor 2
Gene Aliases
-
Chromosome Location
Chr.9: 37181121 - 37207148 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005684672.2 XP_005684729.1 4 - 5 707 74

View Full Product Details