Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRPS28
Entrez Gene ID
102178554
Gene Name
mitochondrial ribosomal protein S28
Gene Aliases
-
Chromosome Location
Chr.14: 38821653 - 38924988 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005689121.3 XP_005689178.1 1 - 2 751 95
GenBank mRNA JO402524.2 - 1 - 2 367 95
KA353409.1 - 2 - 3 579 95

View Full Product Details