Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
METTL23
Entrez Gene ID
102183452
Gene Name
methyltransferase like 23
Gene Aliases
-
Chromosome Location
Chr.19: 54259455 - 54265174 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005694106.3 XP_005694163.1 3 - 4 721 153
XM_013972345.2 XP_013827799.1 2 - 3 305 153
GenBank mRNA JO595594.2 - 3 - 4 694 153

View Full Product Details