Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNS1
Entrez Gene ID
102185479
Gene Name
potassium voltage-gated channel modifier subfamily S member 1
Gene Aliases
-
Chromosome Location
Chr.13: 73001957 - 73007029 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018056903.1 XP_017912392.1 2 - 3 1119 106

View Full Product Details