Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNG3
Entrez Gene ID
102178280
Gene Name
potassium voltage-gated channel modifier subfamily G member 3
Gene Aliases
-
Chromosome Location
Chr.11: 24712907 - 24765837 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018055171.1 XP_017910660.1 1 - 2 1283 93

View Full Product Details