Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RSPO4
Entrez Gene ID
100860773
Gene Name
R-spondin 4
Gene Aliases
-
Chromosome Location
Chr.13: 59419544 - 59456636 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005688332.3 XP_005688389.2 3 - 4 549 89
GenBank mRNA KA310183.1 - 3 - 4 359 89

View Full Product Details