Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LINS1
Entrez Gene ID
102169424
Gene Name
lines homolog 1
Gene Aliases
LINS
Chromosome Location
Chr.21: 5276410 - 5307262 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005694922.2 XP_005694979.1 4 - 5 810 102
XM_005694923.3 XP_005694980.1 4 - 5 1161 102
XM_013972921.2 XP_013828375.1 4 - 5 1140 102
XM_013972923.2 XP_013828377.1 4 - 5 1161 102
XM_013972924.2 XP_013828378.2 4 - 5 1167 102

View Full Product Details