Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CNGA2
Entrez Gene ID
102171585
Gene Name
cyclic nucleotide gated channel alpha 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_017189518: 704946 - 722447 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005700487.2 XP_005700544.1 2 - 3 534 80

View Full Product Details