Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
PRRT4
Entrez Gene ID
102169936
Gene Name
proline rich transmembrane protein 4
Gene Aliases
-
Chromosome Location
Chr.4: 27575657 - 27585645 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018046967.1 XP_017902456.1 3 - 4 956 116

View Full Product Details