Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BAHD1
Entrez Gene ID
102171076
Gene Name
bromo adjacent homology domain containing 1
Gene Aliases
-
Chromosome Location
Chr.10: 66996638 - 67019894 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018054445.1 XP_017909934.1 6 - 7 2330 87
XM_018054446.1 XP_017909935.1 6 - 7 2327 87
XM_018054447.1 XP_017909936.1 6 - 7 2321 87
GenBank mRNA JO558412.2 - 4 - 5 376 87
JT679396.1 - 8 - 8 119

View Full Product Details