Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CASS4
Entrez Gene ID
102170531
Gene Name
Cas scaffold protein family member 4
Gene Aliases
-
Chromosome Location
Chr.13: 58791765 - 58829364 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018057633.1 XP_017913122.1 4 - 5 914 89
XM_018057635.1 XP_017913124.1 5 - 6 1149 89

View Full Product Details