Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
LHX1
Entrez Gene ID
102168572
Gene Name
LIM homeobox 1
Gene Aliases
-
Chromosome Location
Chr.19: 12920702 - 12928184 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018064166.1 XP_017919655.1 2 - 3 1794 70
GenBank mRNA JO428398.1 - 2 - 3 1595 70

View Full Product Details