Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRIM65
Entrez Gene ID
102171278
Gene Name
tripartite motif containing 65
Gene Aliases
-
Chromosome Location
Chr.19: 54953275 - 54958261 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018063783.1 XP_017919272.1 2 - 3 517 101
XM_018063784.1 XP_017919273.1 2 - 3 517 101
GenBank mRNA KA345069.1 - 1 - 2 706 101

View Full Product Details