Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRGPRF
Entrez Gene ID
102189013
Gene Name
MAS related GPR family member F
Gene Aliases
-
Chromosome Location
Chr.29: 47395274 - 47404595 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018043520.1 XP_017899009.1 2 - 3 341 72
GenBank mRNA KA346987.1 - 2 - 3 335 72

View Full Product Details