Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BCHE
Entrez Gene ID
102187168
Gene Name
butyrylcholinesterase
Gene Aliases
-
Chromosome Location
Chr.1: 101031427 - 101110936 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005675363.2 XP_005675420.1 3 - 4 1831 95
GenBank mRNA JT717059.1 - 4 - 5 1777 95

View Full Product Details