Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IL23A
Entrez Gene ID
102190023
Gene Name
interleukin 23 subunit alpha
Gene Aliases
-
Chromosome Location
Chr.5: 56385268 - 56409937 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005680346.3 XP_005680403.1 1 - 2 2654 62
XM_018048070.1 XP_017903559.1 4 - 5 1143 62
GenBank mRNA JT643711.1 - 7 - 8 1931 62

View Full Product Details