Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BIK
Entrez Gene ID
102174730
Gene Name
BCL2 interacting killer
Gene Aliases
-
Chromosome Location
Chr.5: 112857122 - 112876055 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018048868.1 XP_017904357.1 2 - 3 317 71

View Full Product Details