The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005698895.3 | XP_005698952.1 | 2 - 3 | 528 | 117 |
GenBank mRNA | KA326909.1 | - | 2 - 3 | 696 | 117 |