Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLAIN1
Entrez Gene ID
102186221
Gene Name
SLAIN motif family member 1
Gene Aliases
-
Chromosome Location
Chr.12: 33165617 - 33217920 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018056478.1 XP_017911967.1 3 - 4 1203 67
XM_018056479.1 XP_017911968.1 3 - 4 436 67

View Full Product Details