Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
AHNAK2
Entrez Gene ID
102188806
Gene Name
AHNAK nucleoprotein 2
Gene Aliases
-
Chromosome Location
Chr.21: 68761198 - 68794935 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018065939.1 XP_017921428.1 5 - 6 703 93
GenBank mRNA JO646549.2 - 2 - 3 578 93

View Full Product Details