Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PABPC4L
Entrez Gene ID
102170808
Gene Name
poly(A) binding protein cytoplasmic 4 like
Gene Aliases
-
Chromosome Location
Chr.17: 46737715 - 46742887 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018061534.1 XP_017917023.1 1 - 1 1264 74

View Full Product Details