Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHRNA4
Entrez Gene ID
102170991
Gene Name
cholinergic receptor nicotinic alpha 4 subunit
Gene Aliases
-
Chromosome Location
Chr.13: 53566466 - 53578758 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018057546.1 XP_017913035.1 4 - 5 385 58

View Full Product Details