Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NRBF2
Entrez Gene ID
102177130
Gene Name
nuclear receptor binding factor 2
Gene Aliases
-
Chromosome Location
Chr.28: 26296121 - 26324991 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018042540.1 XP_017898029.1 2 - 3 328 79
GenBank mRNA KA322293.1 - 2 - 3 295 79

Gene Symbol
LOC108637522
Entrez Gene ID
108637522
Gene Name
nuclear receptor-binding factor 2-like
Gene Aliases
-
Chromosome Location
Chr.14: 26533910 - 26536091 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018058348.1 XP_017913837.1 2 - 2 272 79

View Full Product Details