Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLEC12B
Entrez Gene ID
102187464
Gene Name
C-type lectin domain family 12 member B
Gene Aliases
-
Chromosome Location
Chr.5: 98370590 - 98375792 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005680857.3 XP_005680914.2 1 - 2 120 91

View Full Product Details