Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC102171908
Entrez Gene ID
102171908
Gene Name
zinc finger protein 30 homolog
Gene Aliases
ZFP30
Chromosome Location
Chr.18: 48481836 - 48504042 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005692356.3 XP_005692413.1 6 - 7 694 114
XM_018063357.1 XP_017918846.1 5 - 6 602 114

View Full Product Details