Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SUSD3
Entrez Gene ID
102171068
Gene Name
sushi domain containing 3
Gene Aliases
-
Chromosome Location
Chr.8: 84192453 - 84216021 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018052498.1 XP_017907987.1 3 - 4 460 68
GenBank mRNA JO366234.2 - 3 - 4 385 68

View Full Product Details