Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NCK2
Entrez Gene ID
102180601
Gene Name
NCK adaptor protein 2
Gene Aliases
-
Chromosome Location
Chr.11: 45493580 - 45612011 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018055323.1 XP_017910812.1 4 - 5 1253 62
XM_018055325.1 XP_017910814.1 2 - 3 958 62

View Full Product Details