Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SHB
Entrez Gene ID
102183872
Gene Name
SH2 domain containing adaptor protein B
Gene Aliases
-
Chromosome Location
Chr.8: 61662606 - 61800316 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005683946.3 XP_005684003.3 2 - 3 1438 107
XM_018052197.1 XP_017907686.1 2 - 3 1438 107
GenBank mRNA JT627875.1 - 7 - 8 1018 107
JT649362.1 - 5 - 6 1192 107
KA335821.1 - 2 - 3 937 107

View Full Product Details