Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DIPK2B
Entrez Gene ID
102191125
Gene Name
divergent protein kinase domain 2B
Gene Aliases
CXHXorf36
Chromosome Location
Chr.X|NW_017189516: 27256701 - 27295754 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018043895.1 XP_017899384.1 2 - 3 622 64

View Full Product Details