Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SAMD15
Entrez Gene ID
102171432
Gene Name
sterile alpha motif domain containing 15
Gene Aliases
-
Chromosome Location
Chr.10: 14492946 - 14503165 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005686163.3 XP_005686220.1 1 - 2 1739 72
GenBank mRNA JT710594.1 - 1 - 2 242 72

View Full Product Details