Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MS4A15
Entrez Gene ID
102175835
Gene Name
membrane spanning 4-domains A15
Gene Aliases
-
Chromosome Location
Chr.29: 36358472 - 36367446 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005699651.2 XP_005699708.1 5 - 6 640 61
XM_005699652.2 XP_005699709.1 4 - 5 517 61

View Full Product Details